Learn More
Tonapofylline, MedChemExpress
Product Code. 16468817
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
Unit Size:
10 miligramos
5 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16468817 5 mg 5 miligramos
16436228 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468817 Supplier Medchem Express Supplier No. HY14873
This item is not returnable. View return policy
MedChemExpress Tonapofylline (BG 9928) is an orally active and selective adenosine A1 receptor antagonist with a Ki of 7.4 nM for human adenosine A1 receptor (hA1), which displays 915-fold selectivity versus human adenosine A2A receptor and 12-fold selectivity versus human adenosine A2B receptor and is used in development for the treatment of heart failure.
Tonapofylline (BG 9928) is an orally active and selective adenosine A1 receptor antagonist with a Ki of 7.4 nM for human adenosine A1 receptor (hA1), which displays 915-fold selectivity versus human adenosine A2A receptor and 12-fold selectivity versus human adenosine A2B receptor and is used in development for the treatment of heart failure.
Specifications
Nombre del producto químico o material Tonapofylline
Forma física Solid
CAS 340021-17-2
Color Blanco
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C22H32N4O4
Para utilizar con (aplicación) Neuroscience-Neuromodulation
Sinónimo BG 9928
Información de solubilidad DMSO : 100 mg/mL (240.09 mM; Need ultrasonic)
SMILES O=C(O)CCC1(CC2)CCC2(C(N3)=NC(N(CCC)C(N4CCC)=O)=C3C4=O)CC1
Peso molecular (g/mol) 416.51
Formula Weight (peso de la fórmula) 416.51
Porcentaje de pureza 96.0%