Learn More
Tomivosertib, MedChemExpress
Código de producto. 16495487
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
100 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16495487 5 mg 5 miligramos
16494206 100 mg 100 miligramos
16479446 50 mg 50 miligramos
16472748 10 mg 10 miligramos
16410499 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16495487 Proveedor Medchem Express N.º de proveedor HY100022
MedChemExpress Tomivosertib (eFT508) is a potent, highly selective, and orally active MNK1 and MNK2 inhibitor, with IC50s of 1-2 nM against both isoforms. Tomivosertib (eFT508) treatment leads to a dose-dependent reduction in eIF4E phosphorylation at serine 209 (IC50=2-16 nM) in tumor cell lines. Tomivosertib (eFT508) also dramatically downregulates PD-L1 protein abundance.
Tomivosertib (eFT508) is a potent, highly selective, and orally active MNK1 and MNK2 inhibitor, with IC50s of 1-2 nM against both isoforms. Tomivosertib (eFT508) treatment leads to a dose-dependent reduction in eIF4E phosphorylation at serine 209 (IC50=2-16 nM) in tumor cell lines. Tomivosertib (eFT508) also dramatically downregulates PD-L1 protein abundance.
Especificaciones
Nombre del producto químico o material Tomivosertib
Forma física Powder
CAS 1849590-01-7
Color Amarillo claro
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C17H20N6O2
Para utilizar con (aplicación) Cancer-Kinase/protease
Sinónimo eFT508
Información de solubilidad DMSO : 4.35 mg/mL (12.78 mM; Need ultrasonic)
SMILES O=C(C1=C(C)C=C(NC2=NC=NC(N)=C2)C(N13)=O)NC43CCCCC4
Peso molecular (g/mol) 340.38
Formula Weight (peso de la fórmula) 340.38
Porcentaje de pureza 98.07%