Learn More
Tocris Bioscience™ M40
Descripción
M40 is a potent, non-selective galanin receptor antagonist (Ki values are 1.82 and 5.1 nM at GAL1 and GAL2 respectively) that inhibits galanin (1-29) (Cat. No. 2696) binding in rat brain in vitro (IC50 = 3 - 15 nM). Attenuates the antidepressant effects of fluoxetine (Cat. No. 0927) and blocks galanin-induced food intake in vivo. Also exhibits weak partial agonist activity at peripheral GAL2 receptors at doses > 100 nM.
Especificaciones
Especificaciones
| Pureza | 0.95 |
| Contenido y almacenamiento | Store at -20°C |
| Cantidad | 1 mg |