missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ HS 014

Description
HS 014 is a potent and selective melanocortin MC4 receptor antagonist (Ki values are 3.16, 108, 54.4 and 694 nM for cloned human MC4, MC1, MC3 and MC5 receptors respectively). Increases food intake in rats and nociception in mice following central administration in vivo. Also inhibits IL-1β-induced Fos expression in the paraventricular hypothalamus.
Specifications
Specifications
| Pureza | 0.95 |
| Contenido y almacenamiento | Store at -20°C |
| Cantidad | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?