Learn More
TMPyP4 tosylate, MedChemExpress
Description
TMPyP4 tosylate (TMP 1363) is a quadruplex-specific ligand, which inhibits the interaction between G-quadruplexes and IGF-1. TMPyP4 tosylate (TMP 1363) is a telomerase inhibitor with antitumor effects in osteosarcoma cell lines.
Specifications
Specifications
| Nombre del producto químico o material | TMPyP4 tosylate |
| CAS | 36951-72-1 |
| Color | Deep Purple |
| Notas de grado de pureza | Research |
| Fórmula molecular | C72H66N8O12S4 |
| Cantidad | 100 mg |
| Sinónimo | TMP 1363 |
| Información de solubilidad | H2O : 25 mg/mL (18.33 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (7.33 mM; Need ultrasonic) |
| SMILES | C[N+]1=CC=C(/C2=C3C=CC(/C(C4=CC=[N+](C)C=C4)=C5C=C/C(N/5)=C(C6=CC=[N+](C)C=C6)/C(C=C/7)=NC7=C(C8=CC=[N+](C)C=C8)/C9=CC=C2N9)=N\3)C=C1.O=S(C%10=CC=C(C)C=C%10)([O-])=O.O=S(C%11=CC=C(C)C=C%11)([O-])=O.O=S(C%12=CC=C(C)C=C%12)([O-])=O.O=S(C%13=CC=C(C)C=C%13)([O-])=O |
| Peso molecular (g/mol) | 1363.6 |
| Show More |