Learn More
Tocris Bioscience™ TMI 1, >98%
GREENER_CHOICE
Código de producto. 30000874
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30000874 10 mg 10 miligramos
30000996 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30000874 Proveedor Tocris Bioscience™ N.º de proveedor 5960/10
Adam 17 (TACE) and MMP inhibitor; orally bioavailable
Adam 17 (TACE) and MMP inhibitor (IC50 values are 3, 4.7, 6.6, 8.4, 12, 26 and 26 nM for MMP-13, MMP-2, MMP-1, ADAM 17, MMP-9, MMP-7 and MMP-14, respectively). Suppresses TNF-α production in an acute LPS-mouse model. Reduces severity score in an in vivo model of rheumatoid arthritis. Displays selective cytotoxicity to tumor cells and cancer stem cells in vitro. Induces tumor apoptosis in a breast cancer in vivo model. Orally bioavailable.
TRUSTED_SUSTAINABILITY
CAS 287403-39-8
Fórmula molecular C17H22N2O5S2
Especificaciones
Nombre del producto químico o material (3S)-4-[[4-(2-Butyn-1-yloxy)phenyl]sulfonyl]-N-hydroxy-2,2-dimethyl-3-thiomorpholinecarboxamide
Cantidad 10 mg
Solubilidad Soluble to 100mM in DMSO and to 20mM in ethanol
Formula Weight (peso de la fórmula) 398.49
Pureza >98%
Peso molecular 398.492