missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, anatase, nanopowder, 99.7% (metals basis)

Código de producto. 17352791
Cambiar vista
Click to view available options
Cantidad:
500 g
Tamaño de la unidad:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17352791 500 g 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17352791 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor 045603.36
Solicitar formato a granel o personalizado

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 1317-70-0 | O2Ti | 79.87 g/mol

Titanium(IV) oxide is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1317-70-0
Fórmula molecular O2Ti
Peso molecular (g/mol) 79.87
Número MDL MFCD00011269,MFCD00210650
Clave InChI GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Sinónimo titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
Nombre IUPAC dioxotitanio
SMILES O=[Ti]=O

Especificaciones

Nombre del producto químico o material Titanium(IV) oxide
Forma física Powder
Cantidad 500 g
Intervalo de porcentaje de ensayo 99.7% (metals basis)
Información de solubilidad Insoluble in water,dilute acids,organic acids,and dilute alkalis.
Formula Weight (peso de la fórmula) 79.9

RUO – Research Use Only

Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.