Learn More
Titanium(IV) 2-ethylhexyloxide, 95%, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar 077107.22
| Cantidad | 100g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
| 1070-10-6 | |
| 564.76 | |
| KTXWGMUMDPYXNN-UHFFFAOYNA-N | |
| 54603245 | |
| [Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-] |
| C32H68O4Ti | |
| MFCD00015264,MFCD00015264 | |
| tyzor tot, 2-ethylhexyl orthotitanate, tetra 2-ethylhexyl titanate, tetra-2-ethylhexyl titanate, tetrakis 2-ethylhexyl titanate, titanic iv acid, tetrakis 2-ethylhexyl ester, 1-hexanol, titanium 4+ salt | |
| titanium(4+) tetrakis(2-ethylhexan-1-olate) |
Especificaciones
| Titanium (IV) 2-ethylhexyloxide | |
| 100g | |
| Ti[OCH2CH(C2H5)(CH2)3CH3]4 | |
| 3692023 | |
| tyzor tot, 2-ethylhexyl orthotitanate, tetra 2-ethylhexyl titanate, tetra-2-ethylhexyl titanate, tetrakis 2-ethylhexyl titanate, titanic iv acid, tetrakis 2-ethylhexyl ester, 1-hexanol, titanium 4+ salt | |
| [Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-] | |
| 564.76 | |
| 564.79 | |
| 1.482 | |
| 0.926 g/mL |
| 1070-10-6 | |
| C32H68O4Ti | |
| MFCD00015264,MFCD00015264 | |
| 71°C (160°F) | |
| KTXWGMUMDPYXNN-UHFFFAOYNA-N | |
| titanium(4+) tetrakis(2-ethylhexan-1-olate) | |
| 54603245 | |
| 95% | |
| Moisture sensitive |