Learn More
Tocris Bioscience™ Tiplaxtinin
Descripción
Tiplaxtinin is a plasminogen activator inhibitor-1 (PAI-1) inhibitor (IC50 = 2.7 μM). Induces vascular smooth muscle cell apoptosis in vitro and reduces carotid artery neointimal formation in a rat vascular injury model. Promotes thrombolysis in rat and dog models of acute arterial thrombosis. Orally available.
Especificaciones
Especificaciones
| Nombre del producto químico o material | α-Oxo-1-(phenylmethyl)-5-[4-(trifluoromethoxy)phenyl]-1H-indole-3-acetic acid |
| CAS | 393105-53-8 |
| Sinónimo | PAI 039, tiplasinin |
| Cantidad | 50 mg |
| Objetivo | Additional Synthetic Protease Inhibitors |
| Fórmula molecular | C24H16F3NO4 |
| Formula Weight (peso de la fórmula) | 439.38 |
| Pureza | >98% |