missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tilivalline, Tilivalline, TRC
Especificaciones
Especificaciones
| Nombre del producto químico o material | Tilivalline |
| CAS | 80279-24-9 |
| Notas de grado de pureza | HPLC |
| Cantidad | 1 mg |
| Fórmula InChI | InChI=1S/C20H19N3O2/c24-17-9-3-6-13-19(17)22-18(16-8-4-10-23(16)20(13)25)14-11-21-15-7-2-1-5-12(14)15/h1-3,5-7,9,11,16,18,21-22,24H,4,8,10H2/t16-,18-/m0/s1 |
| Formula Weight (peso de la fórmula) | 333.1477 |
| Porcentaje de pureza | >95% |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?