Learn More
Tocris Bioscience™ Tilfrinib
Potent and selective breast tumor kinase (Brk) inhibitor
Brand: Tocris Bioscience™ 5579/50
This item is not returnable.
View return policy
Description
Tilfrinib is a potent and selective breast tumor kinase (Brk, also known as tyrosine-protein kinase 6, PTK6) inhibitor (IC50 = 3.15 nM). Exhibits >1000-fold selectivity for Brk over a panel of other kinases. Inhibits proliferation of breast cancer cells in vitro.Specifications
| 3-(9H-Pyrido[2,3-b]indol-4-ylamino)phenol | |
| 1600515-49-8 | |
| 50 mg |