Learn More
Thermo Scientific™ Ticlopidine Hydrochloride
Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines
Marca: Thermo Scientific™ J63971.14
| Cantidad | 25 g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
| 53885-35-1 | |
| 5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride | |
| Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines |
| 300.25g/mol | |
| Powder | |
| White |
Especificaciones
| 25 g | |
| Powder | |
| 53885-35-1 | |
| 300.25g/mol |
| White | |
| Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines | |
| C14H14ClNS·HCl | |
| 5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine hydrochloride |