Descripción
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Especificaciones
Especificaciones
| Tamaño del tapón | GL14, NPT 1/4 in |
| Color del tapón | Negro |
| Material de revestimiento | PTFE-L |
| Tipo de producto | Adaptador de rosca |