Learn More
Thiabendazole, 98+%
Descripción
Thiabendazole, a metal chelator and helminth-specific fumarate reductase inhibitor, is used primarily as an anthelmintic to control infections by helminth organisms. It is also used to control mold and fungal growth in plants. Also used to treat parasitic infections in humans. It is used clinically to treat threadworm, cutaneous larva migrans, visceral larva migrans, and trichinosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in methanol and dimethyl sulfoxide.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Especificaciones
Especificaciones
| Nombre del producto químico o material | Thiabendazole |
| Punto de fusión | 304°C to 305°C |
| Color | Blanco |
| Cantidad | 10 g |
| Número UN | UN3077 |
| Beilstein | 611403 |
| Índice Merck | 14,9289 |
| Información de solubilidad | Soluble in methanol and dimethyl sulfoxide. |
| Formula Weight (peso de la fórmula) | 201.25 |
| Porcentaje de pureza | ≥98% |
| Mostrar más |
Seguridad y manipulación
- Thiabendazole
- Atención
- PELIGRO ACUÁTICO A LARGO PLAZO Categoría 1
- PELIGRO ACUÁTICO AGUDO Categoría 1
- H400-Muy tóxico para los organismos acuáticos.
- H410-Muy tóxico para los organismos acuáticos, con efectos nocivos duraderos.
- P273-Evitar su liberación al medio ambiente.
- P391-Recoger el vertido.
- P501b-Eliminar el contenido/el recipiente en conforme a la reglamentación local/regional/nacional/internacional.
- MIXTURE LIST-Contain: Thiabendazole