Learn More
Thalidomide - linker 2, Tocris Biosciences™
PROTACs (Proteolysis Targeting Chimeras)
Marca: Tocris Bioscience 6468/25
Descripción
Thalidomide 4'-oxyacetamide-PEG4-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.This product has been recently renamed. The previous name for this product was Thalidomide - linker 2
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
| PROTAC Toolbox | |
| 2245697-85-0 | |
| Thalidomide - linker 2 | |
| 25 mg |