Learn More
Thalidomide - linker 1, Tocris Biosciences™
PROTACs (Proteolysis Targeting Chimeras)
Marca: Tocris Bioscience 6467/25
Descripción
Thalidomide 4'-oxyacetamide-PEG3-amine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.This product has been recently renamed. The previous name for this product was Thalidomide - linker 1
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
| 2245697-84-9 | |
| Thalidomide - linker 1 | |
| 25 mg |