Learn More
Tocris Bioscience™ Thalidomide 5'-amine-PEG2-amine
Descripción
Thalidomide 5'-amine-PEG2-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG2 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 5-[(2-(2-(2-Aminoethoxy)ethoxy)ethyl)amino]-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride |
| CAS | 2357110-58-6 |
| Cantidad | 25 mg |
| Objetivo | CRBN |
| Fórmula molecular | C19H24N4O6.HCl |
| Pureza | 0.95 |