Learn More
Tocris Bioscience™ Thalidomide 4'-oxyacetamide-alkylC8-amine
Descripción
Thalidomide 4'-oxyacetamide-alkylC8-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC8 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Thalidomide amide-alkylC8-amine
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | N-(8-Aminooctyl)-2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]oxy]-acetamide hydrochloride |
| CAS | 2415263-07-7 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C23H30N4O6.HCl |
| Pureza | 95% |