Learn More
Tocris Bioscience™ Thalidomide 4'-ether-PEG2-alkyne
Descripción
Thalidomide 4'-ether-PEG2-alkyne is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG2 linker with alkyne terminal ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTACs R&D.
This product has been recently renamed. The previous name for this product was Thalidomide - linker 5
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-(2,6-Dioxo-3-piperidinyl)-4-[2-[2-(2-propyn-1-yloxy)oxy]ethoxy]-1H-isoindole-1,3(2H)dione |
| CAS | 2098487-52-4 |
| Sinónimo | Thalidomide - linker 5 |
| Cantidad | 25 mg |
| Fórmula molecular | C20H20N2O7 |
| Formula Weight (peso de la fórmula) | 400.38 |
| Tipo de producto | Ligand |
| Pureza | 95% |