Learn More
Tocris Bioscience™ Thalidomide 4'-ether-alkylC8-acid
Description
Thalidomide 4'-ether-alkylC8-acid is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC8 linker with a terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Nombre del producto químico o material | 9-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]oxy]nonanoic acid |
| CAS | 2225148-51-4 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C22H26N2O7 |
| Pureza | 0.95 |