Learn More
Tocris Bioscience™ Thalidomide 4'-ether-alkylC6-azide
Descripción
Thalidomide 4-ether-alkylC6-azide is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus an alkylC3 linker with a terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. Please contact us for SD files of our available Degrader Building Blocks.PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-[(6-Azidohexyl)oxy]-2-(2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione |
| CAS | 2411389-65-4 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C19H21N5O5 |
| Pureza | 95% |