Learn More
Tocris Bioscience™ Thalidomide 4'-ether-alkylC5-acid
Descripción
Thalidomide 4'-ether-alkylC5-acid is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus an alkylC5 linker with a terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 6-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]oxy]hexanoic acid |
| CAS | 2087490-48-8 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C19H20N2O7 |
| Pureza | 0.95 |