Learn More
Tocris Bioscience™ Thalidomide 4'-ether-alkylC2-amine
Descripción
Thalidomide 4-ether-alkylC2-amine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus an alkylC2 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. This product has been recently renamed. The previous name for this product was Thalidomide - linker 6 Please contact us for SD files of our available Degrader Building Blocks.PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 4-(2-Aminoethoxy)-2-(2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione hydrochloride |
| CAS | 2341840-99-9 |
| Cantidad | 25 mg |
| Objetivo | Degrader Building Blocks |
| Fórmula molecular | C15H15N3O5.HCl |
| Pureza | 95% |