Learn More
TH287, MedChemExpress
Código de producto. 16447228
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16447228 10 mg 10 miligramos
16433247 50 mg 50 miligramos
16429757 5 mg 5 miligramos
16407206 100 mg 100 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16447228 Proveedor Medchem Express N.º de proveedor HY16965
MedChemExpress TH287 is a potent and selective inhibitor of MTH1, with an IC50 of 0.8 nM. TH287 is highly selective towards MTH1, with no relevant inhibition of MTH2, NUDT5, NUDT12, NUDT14, NUDT16, dCTPase, dUTPase and ITPA at 100 μM. TH287 could act as a chemotherapeutic agent for cancer research.
TH287 is a potent and selective inhibitor of MTH1, with an IC50 of 0.8 nM. TH287 is highly selective towards MTH1, with no relevant inhibition of MTH2, NUDT5, NUDT12, NUDT14, NUDT16, dCTPase, dUTPase and ITPA at 100 μM. TH287 could act as a chemotherapeutic agent for cancer research.
Especificaciones
Nombre del producto químico o material TH287
CAS 1609960-30-6
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C11H10Cl2N4
Cantidad 10 mg
Información de solubilidad DMSO : 100 mg/mL (371.57 mM; Need ultrasonic)
SMILES NC1=NC(C2=CC=CC(Cl)=C2Cl)=CC(NC)=N1
Peso molecular (g/mol) 269.13
Formula Weight (peso de la fórmula) 269.13
Porcentaje de pureza 98.14%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-programmed cell death