Learn More
Tocris Bioscience™ TGX 221
Potent and selective PI 3-kinase β inhibitor
Marca: Tocris Bioscience™ 5832/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TGX 221 is a potent and selective PI 3-kinase β inhibitor (IC50 values are 0.007, 0.1, 3.5 and 5 μM for the β, δ, γ and α isoforms, respectively). Shows >1,000-fold selectively for PI 3-kinase β over a range of other kinases. Inhibits thrombus formation in animal models.Especificaciones
| 7-Methyl-2-(4-morpholinyl)-9-[1-(phenylamino)ethyl]-4H-pyrido[1,2-a]pyrimidin-4-one | |
| 663619-89-4 | |
| 50 mg |