Learn More
TGF-beta 1, 2, 3 Antibody, R&D Systems™
GREENER_CHOICE
Código de producto. 18760638
Change view
Click to view available options
Cantidad:
1 mg
100 μg
25 μg
500 ug
Tamaño de la unidad:
1 miligramo
100 microgramos
25 microgramos
500 microgramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
18760638 100 μg 100 microgramos
18720937 25 μg 25 microgramos
18122111 1 mg 1 miligramo
18109130 500 ug 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 18760638 Proveedor R&D Systems N.º de proveedor MAB1835R100
Mouse Monoclonal Antibody has been used in 2 publications
TGF beta 1/2/3 Monoclonal specifically detects TGF beta 1/2/3 in Multi-Species samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TGF beta 1/2/3
Aplicaciones Neutralization
Clasificación Monoclonal
Clon 1D11R
Conjugado Unconjugated
Dilución Neutralization 0.25-1.25 ug/mL
Formulación Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Alias de gen ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1
Especie del huésped Mouse
Inmunógeno Bovine bone-derived TGF-beta 1 and TGF-beta 2
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7040
Especificidad de la prueba Detects bovine, chicken, mouse, and human TGF-beta in ELISAs and Western blots. It recognizes human TGF-beta 1, TGF-beta 2, and TGF-beta 3.
Especies diana Multi-species
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1