Learn More
Tocris Bioscience™ TFRGAP-NH2
Descripción
TFRGAP-NH2 is a protease-activated receptor 3 (PAR3) peptide agonist. In vitro, it increases the expression of heme oxygenase (HO)-1 in osteoarthritis synovial fibroblasts (OASFs).
Especificaciones
Especificaciones
| Pureza | 0.95 |
| CAS | 1872435-09-0 |
| Contenido y almacenamiento | Store at -20°C |
| Peso molecular | M.W.-observed: 646.75 |
| Cantidad | 1 mg |