Learn More
Tetrahydro-4H-pyran-4-ol, Thermo Scientific™
Product Code. 11926201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11926201 5 g 5g
11916201 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11926201 Supplier Thermo Scientific Acros Supplier No. 189430050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2081-44-9
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key LMYJGUNNJIDROI-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-ol, tetrahydro-4-pyranol, 4-hydroxytetrahydropyran, tetrahydro-4h-pyran-4-ol, tetrahydro-pyran-4-ol, 2h-pyran-4-ol, tetrahydro, 4-hydroxytetrahydropyan, tetrahydropyran-4-ol, 4-hydroxytetrahydro-4h-pyran, 4-hydroxytetrahydro-2h-pyran
PubChem CID 74956
IUPAC Name oxan-4-ol
SMILES C1COCCC1O
Specifications
Density 1.0700g/mL
Boiling Point 87.0°C (15.0 mmHg)
Flash Point 190°C
Refractive Index 1.46
Quantity 5 g
Beilstein 17,II,106
Specific Gravity 1.07
Formula Weight 102.13
Chemical Name or Material Tetrahydro-4H-pyran-4-ol