missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecanoato de testosterona-d3, TRC
Especificaciones
Especificaciones
| Nombre del producto químico o material | Testosterone Undecanoate-d3 |
| Fórmula molecular | C30H45D3O3 |
| Cantidad | 1 mg |
| Sinónimo | (17β)-17-[(1-Oxoundecyl)oxy]androst-4-en-3-one-d3, Andriol-d3, Andriol Testocaps-d3, Nebido-d3, Org 538-d3, Testosterone Undecylate-d3, Undestor-d3 |
| Fórmula InChI | InChI=1S/C30H48O3/c1-4-5-6-7-8-9-10-11-12-28(32)33-27-16-15-25-24-14-13-22-21-23(31)17-19-29(22,2)26(24)18-20-30(25,27)3/h21,24-27H,4-20H2,1-3H3/t24-,25-,26-,27-,29-,30?/m0/s1/i16D2,27D |
| SMILES | O=C1CC[C@@]2(C)C(CC[C@@]([H])([C@@]34[H])[C@]2([H])CCC3([C@](OC(CCCCCCCCCC)=O)([2H])C([2H])([2H])C4)C)=C1 |
| Peso molecular (g/mol) | 459.72 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?