Learn More
Terphenyllin, MedChemExpress
Código de producto. 16464898
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
1 mg
5 mg
10 mg
Tamaño de la unidad:
1 miligramo
10 miligramos
5 miligramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
16464898 10 mg 10 miligramos
16441219 1 mg 1 miligramo
16407657 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 16464898 Proveedor Medchem Express N.º de proveedor HY119821
MedChemExpress Terphenyllin is a naturally abundant p-terphenyl metabolite isolated from the coral derived fungus Aspergillus candidus, has significant α-glucosidase inhibitory activity.
Terphenyllin is a naturally abundant p-terphenyl metabolite isolated from the coral derived fungus Aspergillus candidus, has significant α-glucosidase inhibitory activity.
Especificaciones
Nombre del producto químico o material Terphenyllin
CAS 52452-60-5
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C20H18O5
Cantidad 10 mg
Información de solubilidad DMSO : 83.33 mg/mL (246.28 mM; Need ultrasonic)
SMILES OC1=CC=C(C=C1)C2=C(C(O)=C(C(OC)=C2)C3=CC=C(C=C3)O)OC
Peso molecular (g/mol) 338.35
Formula Weight (peso de la fórmula) 338.35
Porcentaje de pureza 96.72%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation