Learn More
Telithromycin, MedChemExpress
Código de producto. 16428288
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
10 mg
50 mg
Tamaño de la unidad:
10 miligramos
50 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16428288 10 mg 10 miligramos
16414177 50 mg 50 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16428288 Proveedor Medchem Express N.º de proveedor HYA0062
MedChemExpress Telithromycin (HMR3647) is a novel ketolide antibiotic that structurally resembles macrolides.
Telithromycin belongs to the ketolide family that is characterized by a keto group at position 3 of the macrolide ring and is active against bacteria causing community-acquired pneumonia, acute exacerbation of chronic bronchitis, and acute sinusitis. Telithromycin also has similar immunomodulatory effects as macrolides. Telithromycin can be used for the research of respiratory infections including bronchial asthma.
Especificaciones
Nombre del producto químico o material Telithromycin
Forma física Solid
CAS 191114-48-4
Color Off-White
Cantidad 10 mg
Notas de grado de pureza Research
Fórmula molecular C43H65N5O10
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo HMR3647 RU66647
Información de solubilidad DMSO : 100 mg/mL (123.15 mM; Need ultrasonic)
SMILES O[C@H]1[C@]([H])(O[C@@H]([C@](C)(OC)C[C@@H](C)C2=O)[C@@H](C)C([C@@H](C)C(O[C@H](CC)[C@](OC(N3CCCCN4C=C(C5=CC=CN=C5)N=C4)=O)(C)[C@@]3([H])[C@H]2C)=O)=O)O[C@H](C)C[C@@H]1N(C)C
Peso molecular (g/mol) 812.0
Formula Weight (peso de la fórmula) 812.0
Porcentaje de pureza 95.16%