Learn More
Tebanicline dihydrochloride, MedChemExpress
Código de producto. 16493416
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
25 miligramos
5 miligramos
50 miligramos
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
16493416 25 mg 25 miligramos
16478637 5 mg 5 miligramos
16446446 100 mg 100 miligramos
16412177 50 mg 50 miligramos
16406038 10 mg 10 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Código de producto. 16493416 Proveedor Medchem Express N.º de proveedor HY14316A
MedChemExpress Tebanicline dihydrochloride (Ebanicline dihydrochloride) is a nAChR modulator with potent, orally effective analgesic activity. It inhibits the binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM.
Tebanicline dihydrochloride (Ebanicline dihydrochloride) is a nAChR modulator with potent, orally effective analgesic activity. It inhibits the binding of cytisine to α4β2 neuronal nAChRs with a Ki of 37 pM.
Especificaciones
Nombre del producto químico o material Tebanicline dihydrochloride
CAS 209326-19-2
Color Light Yellow
Notas de grado de pureza Research
Fórmula molecular C9H13Cl3N2O
Cantidad 25 mg
Sinónimo Ebanicline dihydrochlorideABT-594 dihydrochloride
Información de solubilidad H2O : 100 mg/mL (368.23 mM; Need ultrasonic) ∣DMSO : ≥ 34 mg/mL (125.20 mM)
SMILES [H]Cl.ClC(C=C1)=NC=C1OC[C@@H]2NCC2.[H]Cl
Peso molecular (g/mol) 271.57
Formula Weight (peso de la fórmula) 271.57
Porcentaje de pureza 98.0%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Neuroscience-Neuromodulation