missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: SRRNSLVGPG AGPGGQRPSL GPVPPLGSRV SFSGLPLAPA RWVAPSYRTE PVPGERWEAA RAQRAME Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 81%.
Dyneins are multi-subunit, high molecular weight ATPases that interact with microtubules to generate force by converting the chemical energy of ATP into the mechanical energy of movement. Cytoplasmic or axonemal Dynein is an approximately 12 subunit complex of two heavy chains, two intermediate chains to anchor Dynein to its cargo, four smaller intermediate chains and several light chains. It performs functions necessary for cell survival such as organelle transport and centrosome assembly. TCTEX1 is a cytoplasmic dynein light chain found in a complex with Na+ CP type X (SCN10A). TCTEX1D4, also called TCTEX2 beta, interacts with different members of the TGF-beta receptor family, and when overexpressed, inhibits TGF-beta signaling.
Especificaciones
Especificaciones
| Antígeno | TCTEX1D4 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | TCTEX1D4 |
| N.º de referencia del gen | Q5JR98 |
| Alias de gen | 4833401D15Rik; LOC102551208; N22.1; novel Tctex-1 family domain-containing protein; protein N22.1; Tctex1 domain containing 4; tctex1 domain-containing protein 4; tctex1 domain-containing protein 4-like; Tctex1d4; Tctex2 beta; Tctex-2-beta |
| Símbolos de los genes | TCTEX1D4 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
