Learn More
TCR γδ Mouse anti-Human, FITC, Clone: B1, BD
Descripción
Recognizes the γ/δ T-cell receptor (TCR). This receptor complex consists of two disulfide-linked glycoproteins, a γ chain (45-60kDa) and a δ subunit (40-60kDa). γ/δ TCR is expressed in less than 10% of human peripheral T cells. The physiological significance of γ/δ+ T cells is still unknown. There is evidence indicating that these cells recognize bacterial ligands and some tumor cells. Reports suggest that γ/δ+ T cells may play a role in the immune reaction during infection and in regulation of pathphysiological autoimmune responses. Due to the fact that CD3 and TCRγδ antibodies may recognize epitopes which are very close to each other and that CD3 was interfering with the clone B1 antibody binding but not with the clone 11F2 antibody, we recommend to use clone 11F2 antibody to perform double staining TCRγδ and CD3. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | TCR γδ |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | B1 |
| Concentración | 0.5mg/mL |
| Conjugado | FITC |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Método de purificación | Affinity Purified |
| Cantidad | 0.1 mg |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.