Learn More
Invitrogen™ TCR delta Monoclonal Antibody (TS-1)
GREENER_CHOICE
Código de producto. 11868821
Change view
Click to view available options
Cantidad:
200 μg
Tamaño de la unidad:
200 microgramos
11868821 200 μg 200 microgramos
1 options
Código de producto. 11868821 Proveedor Invitrogen™ N.º de proveedor TCR1055
Mouse Monoclonal Antibody
TCR1055 targets TCR Delta TCS1 in FACS, IHC(F), IP, and TCA applications and shows reactivity with Human samples. The TCR1055 immunogen is human TCR Vdelta1-Jdelta1. TCR1055 detects TCR Delta TCS1 which has a predicted molecular weight of approximately 17 kDa.
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno TCR delta
Aplicaciones Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation
Clasificación Monoclonal
Clon TS-1
Concentración 0.15 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.5% BSA and 0.1% sodium azide
génica TRDV1
N.º de referencia del gen A0A1B0GX56
Alias de gen CD3 antigen; CD3d antigen; OKT3
Símbolos de los genes TRD, TRDV1
Especie del huésped Mouse
Inmunógeno Human TCR Vdelta1-Jdelta1.
Método de purificación Purified
Cantidad 200 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 28518, 6964
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • TCR delta Monoclonal Antibody (TS-1)
Palabra de advertencia
  • Atención
Clasificación
  • Toxicidad aguda Categoría 4
Indicaciones de peligro
  • H302-Nocivo en caso de ingestión.
Consejos de prudencia
  • P260-No respirar el polvo/el humo/el gas/la niebla/los vapores/el aerosol.
  • P264-Lavarse concienzudamente tras la manipulación.
  • P301+P310-EN CASO DE INGESTIÓN: Llamar inmediatamente a un CENTRO DE TOXICOLOG A/médico/.
  • P501b-Eliminar el contenido/el recipiente en conforme a la reglamentación local/regional/nacional/internacional.
Otros peligros
  • MIXTURE LIST-Contain: sodium azide