missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCR delta Monoclonal Antibody (TS-1), FITC
GREENER_CHOICE

Código de producto. 10699034
Change view
Click to view available options
Cantidad:
200 Tests
Tamaño de la unidad:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
10699034 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10699034 Proveedor Invitrogen™ N.º de proveedor TCR2055

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

TCR2055 targets TCR Delta TCS1 in FACS applications and shows reactivity with Human samples. The TCR2055 immunogen is human TCR Vdelta1-Jdelta2. TCR2055 detects TCR Delta TCS1 which has a predicted molecular weight of approximately 127 kDa. Washing step with 0.1% Tween is necessary to expose antigen.

The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno TCR delta
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon TS-1
Concentración 0.1 mg/mL
Conjugado FITC
Formulación PBS with 0.5% BSA and 0.1% sodium azide
génica TRDV1
N.º de referencia del gen A0A1B0GX56
Alias de gen CD3 antigen; CD3d antigen; OKT3
Símbolos de los genes TRD, TRDV1
Especie del huésped Mouse
Inmunógeno Human TCR Vdelta1-Jdelta2.
Método de purificación Protein A
Cantidad 200 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 28518, 6964
Especies diana Human
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.