Learn More
Tocris Bioscience™ TC-I 2014
Potent TRPM8 antagonist
Marca: Tocris Bioscience™ 5410/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TC-I 2014 is a potent TRPM8 antagonist (IC50 values are 0.8, 3.0 and 4.4 nM for canine, human and rat channels respectively). Exhibits antiallodynic properties in pain models in vivo.Especificaciones
| TC-I 2014 | |
| 3-[7-(Trifluoromethyl)-5-[2-(trifluoromethyl)phenyl]-1H-benzimidazol-2-yl]-1-oxa-2-azaspiro[4.5]dec-2-ene | |
| Soluble to 100mM in DMSO and to 100mM in ethanol | |
| Bioactive Small Molecule |
| 1221349-53-6 | |
| 50 mg | |
| Powder |
Título del producto
Research Use Only. Not For Human or Veterinary Use