Learn More
TC-I 2014, MedChemExpress
Código de producto. 16417087
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
Tamaño de la unidad:
5 miligramos
16417087 5 mg 5 miligramos
1 options
Código de producto. 16417087 Proveedor Medchem Express N.º de proveedor HY110199
MedChemExpress TC-I 2014 (compound 5) is a potent and orally active Benzimidazole-containing transient receptor potential melastatin 8 (TRPM8) antagonist, with IC50 values of 0.8 nM, 3.0 nM and 4.4 nM for canine, human and rat channels respectively. TC-I 2014 exhibits antiallodynic properties in pain models.
TC-I 2014 (compound 5) is a potent and orally active Benzimidazole-containing transient receptor potential melastatin 8 (TRPM8) antagonist, with IC50 values of 0.8 nM, 3.0 nM and 4.4 nM for canine, human and rat channels respectively. TC-I 2014 exhibits antiallodynic properties in pain models.
Especificaciones
Nombre del producto químico o material TC-I 2014
Forma física Solid
CAS 1221349-53-6
Color Blanco
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C23H19F6N3O
Para utilizar con (aplicación) Neuroscience-Neuromodulation
SMILES FC(C1=CC=CC=C1C2=CC(C(F)(F)F)=C3C(N=C(C4=NOC5(CCCCC5)C4)N3)=C2)(F)F
Peso molecular (g/mol) 467.41
Formula Weight (peso de la fórmula) 467.41
Porcentaje de pureza 99.0%