Learn More
Tocris Bioscience™ TC-G 1008
Potent and selective GPR39 agonist
Marca: Tocris Bioscience™ 5355/50
Descripción
Potent and selective GPR39 agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). Selective over a panel of kinases (IC50s > 10 μM) and displays minimal binding affinity for ghrelin and neurotensin-1 receptors (IC50s > 30 μM). Increases GLP-1 levels in vitro and in vivo. Reduces TNF-α-induced oxidative stress, mitochondrial dysfunction, cytokine expression and secretion of MMPs in fibroblast-like synoviocytes (FLS). Orally bioavailable.
TC-G 1008 is a potent and selective GPR39 agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). Selective over a panel of kinases (IC50s > 10 μM) and displays minimal binding affinity for ghrelin and neurotensin-1 receptors (IC50s > 30 μM). Increases GLP-1 levels in vitro and in vivo. Reduces TNF-α-induced oxidative stress, mitochondrial dysfunction, cytokine expression and secretion of MMPs in fibroblast-like synoviocytes (FLS). Orally bioavailable.Especificaciones
| TC-G 1008 | |
| N-[3-Chloro-4-[[[2-(methylamino)-6-(2-pyridinyl)-4-pyrimidinyl]amino]methyl]phenyl]methanesulfonate | |
| Soluble to 20mM in DMSO | |
| Bioactive Small Molecule |
| 1621175-65-2 | |
| 50 mg | |
| Solid |
Research Use Only. Not For Human or Veterinary Use