Learn More
Tocris Bioscience™ TC-G 1005
Potent and selective GPBA receptor (TGR5) agonist
Marca: Tocris Bioscience™ 5129/10
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TC-G 1005 is a potent and selective GPBA agonist (EC50 = 0.72 nM for hTGR5). Selective for TGR5 over FXR (farnesoid X receptor). Increases plasma GLP-1 levels and reduces blood glucose in mice. Orally available.Especificaciones
| TC-G 1005 | |
| (4-Cyclopropyl-3,4-dihydro-1(2H)-quinoxalinyl)[4-(2,5-dimethylphenoxy)-3-pyridinyl]methanone | |
| Soluble to 100mM in DMSO and to 100mM in ethanol | |
| Bioactive Small Molecule |
| 1415407-60-1 | |
| 10 mg | |
| Solid |
Título del producto
Research Use Only. Not For Human or Veterinary Use