missing translation for 'onlineSavingsMsg'
Learn More

TAU Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1E9A8, Proteintech

Código de producto. 16828435 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16828435 20 μL 20 microlitros
16818435 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16828435 Proveedor Proteintech N.º de proveedor 664991IG20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Tau is a neuronal microtubule-associated protein found predomitly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer′s diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclindependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick′s disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.

Especificaciones

Antígeno TAU
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 1E9A8
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica MAPT
N.º de referencia del gen P10636, P10637, P19332
Alias de gen DDPAC, FTDP 17, MAPT, MAPTL, MSTD, MTBT1, MTBT2, Neurofibrillary tangle protein, Paired helical filament tau, PHF tau, PPND, TAU
Símbolos de los genes Mapt
Especie del huésped Mouse
Inmunógeno TAU Fusion Protein Ag21926
Método de purificación Protein A
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 100738351, 17762, 29477, 4137
Especies diana Human, Mouse, Rat, Pig
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2c
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.