Learn More
MedChemExpress Tat-NR2B9c TFA
Descripción
Tat-NR2B9c TFA disrupts the PSD-95/NMDAR interaction, inhibiting NR2A and NR2B binding to PSD-95 with IC50 values of 0.5 μM and 8 μM, respectively. Tat-NR2B9c TFA also inhibits neuronal nitric oxide synthase (nNOS)/PSD-95 interaction, and possesses neuroprotective efficacy.
Especificaciones
Especificaciones
| Pureza | 98.99% |
| CAS | 1834571-04-8 |
| Contenido y almacenamiento | Protect from light, stored under nitrogen∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen) |
| Para utilizar con (aplicación) | Neuroscience-Neurodegeneration |
| Peso molecular | 2632.9 g/mol |
| Tipo de producto | Peptides |
| Cantidad | 5 mg |
| Condiciones de envío | Room temperature |
| Solubilidad | H2O : ≥ 50 mg/mL (18.99 mM) |
| Sinónimo | Tat-NR2Bct TFA∣NA-1 TFA |