Learn More
Tocris Bioscience™ TAT-cyclo-CLLFVY
Descripción
TAT-cyclo-CLLFVY is a selective HIF-1 dimerization inhibitor. Blocks protein-protein interaction of recombinant HIF-1α, but not HIF-2α, with HIF-1β (IC50 = 1.3 μM) . Inhibits hypoxia-induced HIF-1 activity, and decreases VEGF and CAIX expression in osteosarcoma and breast cancer cells in vitro. Also reduces tubularization of hypoxic HUVECs.
Especificaciones
Especificaciones
| Pureza | 95% |
| CAS | 1446322-66-2 |
| Contenido y almacenamiento | Store at -20°C |
| Descripción | HIF Inhibitors |
| Peso molecular | Observed M.W.: 2559.1 |
| Cantidad | 1 mg |
| Forma estructural | C111H188N42O24S2 |