Learn More
Tocris Bioscience™ TASP 0390325
Potent and selective V1B receptor antagonist; orally bioavailable
Marca: Tocris Bioscience™ 5729/50
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TASP 0390325 is a potent and selective V1B receptor antagonist (IC50 = 2.22 nM for rat pituitary receptor). Exhibits no significant affinity for 85 other receptors, ion channels and transporters at 10 μM, including V1A and V2. Increases ACTH levels in rats. Orally bioavailable. Antidepressant and anxiolytic.Especificaciones
| Potent and Selective V1B Receptor Antagonist, Orally Bioavailable | |
| 1642187-96-9 | |
| 554.44g/mol | |
| Vasopressin Receptor Antagonists |
| Vasopressin Receptor | |
| C25H29ClFN5O4.HCl | |
| Store at -20°C | |
| 50 mg |
Título del producto
For Research Use Only