missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R3 Polyclonal Antibody, Invitrogen™
Descripción
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μL of sterile water
This gene encodes a member of a family of candidate taste receptors that are members of the G protein-coupled receptor superfamily and that are specifically expressed by taste receptor cells of the tongue and palate epithelia. These apparently intronless taste receptor genes encode a 7-transmembrane receptor protein, functioning as a bitter taste receptor. This gene is clustered with another 3 candidate taste receptor genes in chromosome 7 and is genetically linked to loci that influence bitter perception. Gustducin-coupled receptor implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in subsets of taste receptor cells of the tongue and palate epithelium and exclusively in gustducin-positive cells. Expressed in the antrum and fundus (part of the stomach), duodenum and in gastric endocrine cells. Several bitter taste receptors are expressed in a single taste receptor cell.
Especificaciones
Especificaciones
| Antígeno | TAS2R3 |
| Aplicaciones | Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| génica | TAS2R3 |
| N.º de referencia del gen | Q9NYW6 |
| Alias de gen | Taste receptor type 2 member 3, T2R3, TAS2R3 |
| Símbolos de los genes | TAS2R3 |
| Especie del huésped | Rabbit |
| Inmunógeno | Synthetic peptide from human taste receptor, type 2, member 3. |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?