Learn More
Taranabant, MedChemExpress
Código de producto. 16495527
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
2 mg
5 mg
10 mg
25 mg
Tamaño de la unidad:
10 miligramos
2 miligramos
25 miligramos
5 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16495527 5 mg 5 miligramos
16479995 2 mg 2 miligramos
16462788 10 mg 10 miligramos
16472526 25 mg 25 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16495527 Proveedor Medchem Express N.º de proveedor HY10013
MedChemExpress Taranabant is a highly potent and selective cannabinoid 1 (CB1) receptor inverse agonist that inhibits the binding and functional activity of various agonists, with a binding Ki of 0.13 nM for the human CB1R in vitro.
Taranabant is a highly potent and selective cannabinoid 1 (CB1) receptor inverse agonist that inhibits the binding and functional activity of various agonists, with a binding Ki of 0.13 nM for the human CB1R in vitro.
Especificaciones
Nombre del producto químico o material Taranabant
Forma física Solid
CAS 701977-09-5
Color Off-White
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C27H25ClF3N3O2
Para utilizar con (aplicación) Metabolism-protein/nucleotide metabolism
Sinónimo MK-0364
Información de solubilidad DMSO : ≥ 42 mg/mL (81.40 mM)
SMILES ClC1=CC=C(C[C@@H](C2=CC=CC(C#N)=C2)[C@@H](NC(C(C)(OC3=CC=C(C=N3)C(F)(F)F)C)=O)C)C=C1
Peso molecular (g/mol) 515.95
Formula Weight (peso de la fórmula) 515.95
Porcentaje de pureza 99.03%