Learn More
Tocris Bioscience™ TAPI 2
Descripción
TAPI 2 is an ADAM-17 (TACE) and MMP inhibitor (Ki = 120 nM at ADAM-17). Sensitizes cancer stem cells to the lethal effects of 5-FU in vitro. Blocks shedding of TNF-α from cell membranes.
Especificaciones
Especificaciones
| Pureza | >95% |
| CAS | 689284-12-6 |
| Contenido y almacenamiento | Store at -20°C |
| Descripción | N-[(2R)-2-[2-(Hydroxyamino)-2-oxoethyl]-4-methyl-1-oxopentyl]-3-methyl-L-valyl-N-(2-aminoethyl)-L-alaninamide |
| Peso molecular | 415.54 |
| Cantidad | 1 mg |