Learn More
Tocris Bioscience™ TAPI 1
Descripción
TAPI 1 is a TACE/ADAM-17 and MMP inhibitor. Blocks shedding of TNF from cell membranes. Reduces pain-associated behavior in mice with a constructive mononeuropathy.
This product is typically reconstituted in DMSO.
Especificaciones
Especificaciones
| Pureza | >95% |
| CAS | 163847-77-6 |
| Contenido y almacenamiento | Store at -20°C |
| Descripción | N-[(2R)-2-[2-(Hydroxyamino)-2-oxoethyl]-4-methyl-1-oxopentyl]-3-(2-naphthalenyl)--alanyl-L-alaninamide |
| Peso molecular | 499.6 |
| Cantidad | 1 mg |