Learn More
Tocris Bioscience™ TAPI 0
Descripción
TAPI 0 is an ADAM-17 (TACE) and MMP inhibitor; attenuates TNF-α processing. Acts in concert with GM6001 (Cat. No. 2983) to inhibit Chlamydia trachomatis growth.
Especificaciones
Especificaciones
| Pureza | >95% |
| CAS | 163958-73-4 |
| Contenido y almacenamiento | Store at -20°C |
| Descripción | N-[(2R)-2-[2-(Hydroxyamino)-2-oxoethyl]-4-methyl-1-oxopentyl]-3-(2-naphthalenyl)-L-alanyl-L-alaninamide |
| Peso molecular | 456.54 |
| Cantidad | 1 mg |