Learn More
Tank Liner, Aegis™ 5-14, top and bottom drain
GREENER_CHOICE
Código de producto. 17918281 Tienda Thermo Scientific Productos
Change view
Click to view available options
Capacidad (métrico):
50 L
100 L
200 L
Tamaño de la unidad:
paquete de 10
paquete de 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Capacidad (métrico) unitSize
17918281 100 L paquete de 5
17928281 50 L paquete de 10
17948241 200 L paquete de 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 17918281 Proveedor Thermo Scientific N.º de proveedor SH31306.02
Use Thermo Scientific Tank Liners for non-sterile fluid handling operations such as waste collection or powdered media or buffer hydration processes. Tank Liners are constructed with our Thermo Scientific Aegis 5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility.
Features include:
• Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
• Gamma-irradiation sterilized
• Squared tube design for best fit and reliability
• Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
• Customizable via the industry's largest library of qualified components
Recommended for:
• Harvest—collection and storage of bulk non-sterile process liquids
• Collect—non-sterile process liquids for further processing
• Hydrate—powdered buffers, media, and media supplements
TRUSTED_SUSTAINABILITY
Especificaciones
Capacidad (métrico) 100 L
Para utilizar con (aplicación) Single-Use Bioprocessing
Cantidad 100 L
Tubing Style TPE